missing translation for 'onlineSavingsMsg'
Learn More
Learn More
behr Labor-Technik™ HDPE, Canister with Screw Cap and UN Approval
Description
- White High-Density Polyethylene canisters
- UN-certified
- Screw-on cap
- Suitable for hazardous goods
Specifications
Specifications
| Capacity (English) | 6.6 gal. |
| Capacity (Metric) | 25 L |
| Certifications/Compliance | UN-certified |
| Closure Color | Black |
| Closure Material | Polyethylene |
| Closure Type | Screw Cap |
| Color | White |
| Description | PE canister 25 L, white |
| For Use With (Application) | For Transport and Storage of Hazardous Goods |
| Includes | Screw cap |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?