Learn More
Macherey Nagel Capillary column OPTIMA-17 MSID: 0.25 mm, film thickness: 0.25 µm length: 60 m
Brand:  Macherey Nagel 726162.60
Capillary column OPTIMA-17 MSID: 0.25 mm, film thickness: 0.25 µm length: 60 m
Product Code. 12753371
  • 1180.00€ / Pack of 1
Estimated Shipment: 25-09-2026