Learn More
Trajan Scientific and Medical™ Capillary Electrophoresis Tubing
Ideal for field coating applications.
Brand:  Trajan Scientific and Medical™ 062801
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15278843
  • 50.75€ / 1m
Estimated Shipment: 13-05-2024
to see stock.
Add to basket
Description
Description
  • Manufactured from precision bore tubing
  • Wide pH range
  • 30, 50, 75 and 100μm ID
  • Column-to-column reproducibility
  • Untreated capillary electrophoresis tubing
Specifications
Specifications
Electrophoresis Tubing Untreated 0.363mm OD x 0.03mm ID x 1m Length
Hydrophobic and hydrophilic surface treatments
0.03 mm
1 m
Untreated
0.001 in.
0.363 mm
Precision Bore Tubing