Learn More
Thermo Scientific™ Thermo Scientific™ Specimen Collection Beakers
GREENER_CHOICE
Product Code. 15638558
Click to view available options
Capacity (Metric):
103.5 mL
236 mL
Capacity (English):
3.5 oz.
8 oz.
Includes:
Pour Spout
Temperature Strip
Unit Size:
Pack of 350
Pack of 500
Product Code. 15638558
Brand: Thermo Scientific™ 10126
Thermo Scientific Collection Cups are used for for traditional and on-site testing.
Thermo Scientific™ Collection Beakers are offered in two volumes, 3 oz. and 8 oz., to support a variety of sample collection needs. The beakers feature pour spouts for easy sample transfer.
Features:
  • Pour spout for easy transfer of sample into smaller containers
  • With or without temperature indicator strips
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specimen Collection Cup
Material Polypropylene
Capacity (English) 8 oz.
Capacity (Metric) 236 mL
Description Collection Cup with temperature strip; 8 oz. (236 mL)
Includes Temperature Strip
No. per Case 500/Cs.