Learn More
CAPSO, 0.2M buffer soln., pH 9.0, Thermo Scientific™
Product Code. p-7042034
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
Product Code. 15473769
Brand: Thermo Scientific Alfa Aesar J61126.AE
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 102601-34-3
Concentration 0.2 M
pH 9.0
Physical Form Liquid
Quantity 100 mL
Molecular Formula C9H12NO4S
Odor Odorless
MDL Number MFCD00070063
InChI Key CLEVULOKVPBHCU-VIFPVBQESA-M
SMILES O[C@@H](CNC1=CC=CC=C1)CS([O-])(=O)=O
IUPAC Name (2S)-2-hydroxy-3-(phenylamino)propane-1-sulfonate
Molecular Weight (g/mol) 230.26