Learn More
Erlab™ Captair™ Mobile Ductless Fume Hood Filters
Provides personal protection while handling liquid, gaseous and solid chemicals. Erlab™ Captair™ Mobile Ductless Fume Hood Filters provide filtration of organic, acid, formadehyde, and ammonia vapors as well as fine powders.
Brand: Erlab™ H01074301
Additional Details : Weight : 1.00000kg
Description
Provides personal protection during handlings of liquid, gaseous and solid chemicals.Filter types available:
- GF4 AS: Oganic vapors
- GF4 BE +: Organix vapors plus acid vapors
- GF4 F: Formaldehyde vapors
- GF4 K: Ammonia vapors
- GF4 HP: HI4 HEPA filter for powders
Specifications
K | |
Carbon Filter | |
For Ammonia Vapors |
834/1634 Mobile Ductless Filtration Fume Hood | |
Replacement Filter |