Learn More
Erlab™ CaptairStore™ and ChemTrap™ Filters
Replaces filters in CaptairStore™ and ChemTrap™ filtration systems. CaptiarStore™ and ChemTrap™ Filters are available in a variety of models designed and optimized for specific filtration applications.
Brand: Erlab™ H11112201
Additional Details : Weight : 0.01000kg
Description
Replacement and spare filters for CaptiarStore™ and ChemTrap™ filtration systems.ChemTrap™ units have been designed to vent and filter vapours coming from chemicals stored in fireproof safety cabinets. They allow fireproof safety cabinets to be totally autonomous by avoiding any connection to energy consuming air extraction systems.
CaptairStore™ filtered storage cabinets are an ideal solution for any laboratory where many flasks and bottles containing toxic or noxious chemicals pollute the lab air and take up valuable bench space.
Specifications
BE | |
Captair™ Store™ 832 and ChemTrap™ H402 | |
Spare Filter | |
Polyvalent for Acid and Organic Vapors |