Learn More
Erlab™ Captair™ Work Surfaces
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Work Surfaces are for use with Captair PCR work stations.
Brand: Erlab™ I0120030001
Additional Details : Weight : 1.00000kg
Description
- High chemical and mechanical resistance
- Rounded corners to facilitate cleaning operations
- Built in spill tray
Specifications
For Model XLS71 Laboratory Furniture | |
Table Top | |
Ideal for Precision Weighing |
Sealed Frame | |
Phenolic Resin |