missing translation for 'onlineSavingsMsg'
Learn More

Captopril, Thermo Scientific™

Orally active inhibitor of angiotensin-converting enzyme

Brand:  Thermo Scientific Alfa Aesar J63593.06

Quantity 5g

 View more versions of this product

Product Code. 15445789

  • 189.00€ / 5g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Captopril has also been shown to inhibit the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density. Captopril demonstrates noncompetitive inhibition of tyrosinase monophenolase activity and competitive inhibition of diphenolase activity

Solubility
Soluble in water (160 mg/ml) at 25°C, sesame and corn oils (<1mg/mL) at 25°C, methanol (>100 mg/ml), ethanol (>100 mg/ml), isopropanol (>100 mg/ml), chloroform (>100 mg/ml), and dichloromethane (>100 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

62571-86-2
217.283
FAKRSMQSSFJEIM-RQJHMYQMSA-N
44093
(2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
C9H15NO3S
MFCD00168073
captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril
CHEBI:3380
CC(CS)C(=O)N1CCCC1C(=O)O

Specifications

62571-86-2
MFCD00168073
477887
captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril
FAKRSMQSSFJEIM-RQJHMYQMSA-N
(2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
44093
217.29
C9H15NO3S
5g
14,1774
Soluble in water (160mg/ml) at 25°C,sesame and corn oils (<1mg/ml) at 25°C,methanol (>100mg/ml),ethanol (>100mg/ml),isopropanol (>100mg/ml),chloroform (>100mg/ml),and dichloromethane (>100mg/ml).
CC(CS)C(=O)N1CCCC1C(=O)O
217.283
CHEBI:3380
Captopril
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.