missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Alexa Fluor™ 647 Anti-AAV9 Conjugate

Product Code. 30484879 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 30484879

Brand: Thermo Scientific™ 7233332100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Alexa Fluor™ 647 Anti-AAV9 Conjugate contains a 13-kDa recombinant single-domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity to adeno-associated virus particles type 9 (AAV9).

CaptureSelect Alexa Fluor 647 Anti-AAV9 Conjugate contains a 13-kDa recombinant single-domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity to adeno-associated virus particles type 9 (AAV9). This anti-AAV9 affinity ligand is chemically conjugated to Alexa Fluor 647 (AF647) via an appropriate spacer that fully retains the binding reactivity of the ligand, enabling detection of AAV9 total capsids.

Conjugation of the CaptureSelect AAV9 affinity ligand with the AF647 dye combines the high binding affinity of the ligand with the unique characteristics of AF647 dye, which is a bright, far-red–fluorescent dye with excitation for the 594- or 633-nm laser lines. This combination makes this product ideally suited for stable signal generation in fluorescent immuno-assay (FIA) applications and imaging.

This conjugate and our other anti-AAV9 conjugates, including CaptureSelect Biotin Anti-AAV9 and CaptureSelect HRP Anti-AAV9 (Cat. No. 7103332100 and 7303332100), bind with high affinity to AAV capsids of the AAV9 serotype. For the detection of other AAV serotype capsids we recommend CaptureSelect Biotin Anti-AAVX, CaptureSelect HRP Anti-AAVX, and CaptureSelect AF647 Anti-AAVX conjugates (Cat. No. 7103522100, 71033522500, 7303522100, 7233522100).

Product characteristics

  • Format type: Alexa Fluor, liquid
  • Buffer: PBS
  • Storage: store in freezer (-5 to -30°C)
  • Concentration: 0.5 mg/mL
  • Volume: 0.2 mL
  • Working dilution: the optimal working dilution should be determined empirically for each application

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin Free).

Specifications

Capacity (Metric) 200 μL
Concentration 0.5 mg/mL
Content And Storage Store in freezer (-5 to -30°C).
Form Liquid
Product Line CaptureSelect
Quantity 100 μg
For Use With (Application) Detection
Target AAV9 serotype
Label or Dye Alexa Fluor™ 647
Ligand Type AAV9 Affinity Ligand

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.