missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbamic Acid Propyl Ester, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | n-Propylcarbamate |
| CAS | 627-12-3 |
| Molecular Formula | C4 H9 N O2 |
| Quantity | 100 mg |
| Synonym | Carbamic acid, propyl ester, Propyl carbamate, NSC 4855, n-Propyl carbamate, n-Propylcarbamate, Carbamic acid propyl ester |
| InChI Formula | InChI=1S/C4H9NO2/c1-2-3-7-4(5)6/h2-3H2,1H3,(H2,5,6) |
| SMILES | CCCOC(=O)N |
| IUPAC Name | propyl carbamate |
| Molecular Weight (g/mol) | 103.12 |
| Formula Weight | 103.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction