missing translation for 'onlineSavingsMsg'
Learn More

Carbon disulfide, ACS, 99.9+%, Thermo Scientific Chemicals

Catalog Number p-4408231
Click to view available options
Quantity:
500 mL
2 L
4 x 500 mL
This item is not returnable. View return policy
75-15-0
CS2
76.131
MFCD00011321
QGJOPFRUJISHPQ-UHFFFAOYSA-N
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
6348
CHEBI:23012
methanedithione
C(=S)=S
This item is not returnable. View return policy
75-15-0
CS2
76.131
MFCD00011321
QGJOPFRUJISHPQ-UHFFFAOYSA-N
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
6348
CHEBI:23012
methanedithione
C(=S)=S

For Use in Electronic Vacuum Tubes, in the Manufacturing of Carbon Tetrachloride, Rayon

It is used in electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent, manufacture of rayon, cellophane, and carbon tetrachloride, to produce rubber and chemicals. It is used in the manufacture of xanthates. It is also used as a reaction agent for the production of rubber vulcanization. accelerators and soil sterilizers. It is an effective solvent for oils, waxes, sulfur and sulfur compounds. Carbon disulfide and carbon dioxide acts as resources for functional polymers. It is also useful for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Color (APHA) 10, Evaporation Residue 0.002%, Hydrogen sulfide (H2S) P.T. (limit about 1.5ppm, Sulfur dioxide (SO2) P.T. (limit about 2.5ppm), H2O 0.05%
TRUSTED_SUSTAINABILITY
Melting Point -112°C
Color Colorless
Density 1.2632
Boiling Point 46°C
Quantity 500 mL
Flash Point -30°C (-22°F)
Physical Form Liquid
Refractive Index 1.627
Assay Percent Range 99.9+%
Solubility Information Soluble in alcohol,ether,benzene,oil,chloroform and carbon tetrachloride.
Formula Weight 76.14
Grade ACS Reagent
Chemical Name or Material Carbon disulfide, For analysis APS >5 micron
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'