missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, AISI 1018, G10180, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Carbon Steel, AISI 1018, G10180, Ø40x20mm
Specifications
Specifications
| Chemical Name or Material | Zinc/Tin/Arsenic/Molybdenum/Nitrogen/Phosphorus/Aluminium/Nickel/Copper/Chromium/Silicon/Carbon/Manganese |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Zinc 0.0005 %/Tin 0.0024 %/Arsenic 0.0029 %/Molybdenum 0.0047 %/Nitrogen 0.0052 %/Phosphorus 0.015 %/Aluminium 0.0198 %/Nickel 0.0284 %/Copper 0.05 %/Chromium 0.0793 %/Silicon 0.147 %/Carbon 0.171 %/Manganese 0.803 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction