Learn More
Swann-Morton™ Carbon Steel Scalpel Blades
Click to view available options
Size:
# 14
# 20
# 22A
# 27
# 36
60 x 10.96 mm
Major-Z
Unit Size:
Pack of 100
Pack of 50
Description
Use Swann Morton™ Carbon Steel Scalpel Blades in the operating room for precise cutting and repair.
- Carbon steel to BS EN 27740
- Handles and blades sold separately
Specifications
Specifications
| Blade Material | Carbon Steel |
| Product Type | Scalpel Blade |
| Size | # 22A |
| Description | Surgeon Grade |
| Sterility | Non-sterile |
| Quantity | 10/Pk. |