Learn More
Alfa Aesar™ Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Brand: Alfa Aesar™ 044955.06
Additional Details : CAS : 16971-33-8 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
16971-33-8 | |
954.429 | |
ITDZWCWUTIWRCJ-UHFFFAOYSA-M | |
71463878 | |
[H-].C=O.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Ru+2] |
C55H48ClOP3Ru | |
MFCD00049804 | |
carbonylchlorohydridotris triphenylphosphine-ruthenium ii | |
formaldehyde;hydride;ruthenium(2+);triphenylphosphane;chloride |
Specifications
Carbonylchlorohydridotris (triphenylphosphine)ruthenium (II) | |
Gray | |
5g | |
C55H48ClOP3Ru | |
MFCD00049804 | |
Insoluble in water,ethanol,and DMSO. Sparingly soluble in benzene,chloroform and acetone. | |
[H-].C=O.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Ru+2] | |
954.429 | |
952.41 |
Light sensitive; Very Hygroscopic | |
204°C to 206°C (decomposition) | |
16971-33-8 | |
[(C6H5)3P]3Ru(CO)(Cl)H | |
carbonylchlorohydridotris triphenylphosphine-ruthenium ii | |
ITDZWCWUTIWRCJ-UHFFFAOYSA-M | |
formaldehyde;hydride;ruthenium(2+);triphenylphosphane;chloride | |
71463878 | |
Crystalline |