Learn More
Novus Biologicals™ Carboxyl IR Fluorescent Particles, UltraBlue, 3.5-3.9 um
Shop All Bio Techne Products
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
Specifications
Specifications
| Content And Storage | Store at 4°C. Do not freeze. |
| Color | UltraBlue |
| For Use With (Application) | Flow Cytometry |
| Material | Polystyrene |
| Product Type | Fluroescent Carboxylate-Modified Particle |
| Quantity | 2 mL |
| Diameter (Metric) | 3.5 to 3.9 μm |