missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethylcellulose sodium salt, Thermo Scientific Chemicals

Catalog Number p-7023991
Click to view available options
Quantity:
500 g
2500 g
9004-32-4
(C12 H14 O9 R6)n
263.20
MFCD00081472
DPXJVFZANSGRMM-UHFFFAOYNA-N
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
23706213
sodium;2,3,4,5,6-pentahydroxyhexanal;acetate
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
9004-32-4
(C12 H14 O9 R6)n
263.20
MFCD00081472
DPXJVFZANSGRMM-UHFFFAOYNA-N
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
23706213
sodium;2,3,4,5,6-pentahydroxyhexanal;acetate
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O

Carboxymethylcellulose sodium salt is used in drilling muds, in detergents as a soil-suspending agent, in resin emulsion paints, adhesives, printing inks, textile sizes and protective colloid. It acts as a stabilizer in foods. It is also employed in pharmaceuticals as a suspending agent and excipients for tablets. It is used as viscosity modifiers to stabilize the emulsions. It is used as a lubricant in artificial tears and it is used to characterize enzyme activity from endoglucanases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carboxymethylcellulose sodium salt is used in drilling muds, in detergents as a soil-suspending agent, in resin emulsion paints, adhesives, printing inks, textile sizes and protective colloid. It acts as a stabilizer in foods. It is also employed in pharmaceuticals as a suspending agent and excipients for tablets. It is used as viscosity modifiers to stabilize the emulsions. It is used as a lubricant in artificial tears and it is used to characterize enzyme activity from endoglucanases.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 500 g
Merck Index 14,1829
Solubility Information Soluble in water.
Chemical Name or Material Carboxymethylcellulose sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'