Learn More
Careez Solution(II), Chem Lab™
Click to view available options
Quantity:
1 L
Specifications
Color | Yellow |
Boiling Point | 100°C |
Melting Point | 0°C |
Composition | 106 g K4(Fe(CN)6).3H2O / l H2O |
Density | 1.12g/mL |
Solubility Information | Water Soluble |
Odor | Odorless |
pH | ±9.5 |
Quantity | 1 L |
Physical Form | Liquid |