missing translation for 'onlineSavingsMsg'
Learn More

Carisoprodol, TRC

Product Code. 30137426
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137426 50 mg 50mg
30136987 100 mg 100mg
17974795 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137426 Supplier Toronto Research Chemicals Supplier No. TRCC18358050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-44-4
Formula Weight 260.1736
InChI Formula InChI=1S/C12H24N2O4/c1-5-6-12(4,7-17-10(13)15)8-18-11(16)14-9(2)3/h9H,5-8H2,1-4H3,(H2,13,15)(H,14,16)
IUPAC Name [2-(carbamoyloxymethyl)-2-methylpentyl] N-propan-2-ylcarbamate
Molecular Formula C12 H24 N2 O4
Molecular Weight (g/mol) 260.33
Percent Purity >95%
SMILES CCCC(C)(COC(=O)N)COC(=O)NC(C)C
Synonym Carisoprodol, (2RS)-2-[(Carbamoyloxy)methyl]-2-methylpentyl (1-methylethyl)carbamate, Carbamic acid, N-(1-methylethyl)-, 2-[[(aminocarbonyl)oxy]methyl]-2-methylpentyl ester, Carbamic acid, (1-methylethyl)-, 2-[[(aminocarbonyl)oxy]methyl]-2-methylpentyl ester (9CI), Carbamic acid, isopropyl-, 2-(hydroxymethyl)-2-methylpentyl ester carbamate (ester) (8CI), Carbamic acid, isopropyl-, 2-(hydroxymethyl)-2-methylpentyl ester, carbamate (6CI), 2-Methyl-2-propyl-1,3-propanediol carbamate isopropylcarbamate, Apesan, Arusal, Atonalyt, Calenfa, Caprodat, Carisol, Carisoma, Carisoprodate, Carisoprodatum, Domarax, Flexal, Flexartal, Isobamate, Isomeprobamate, Isopropyl meprobamate, Isoprotan, Isoprotane, Isoprothane, Izoprotan, Miolisodal, Mioril, N-Isopropyl-2-methyl-2-propyl-1,3-propanediol dicarbamate, NIH 10966, NSC 172124, Rela, Relasom, Sanoma, Skutamil, Soma, Somadril, Somalgit, Stialgin
Chemical Name or Material Carisoprodol
Grade HPLC
Show More Show Less

Specifications

Quantity 50 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.