Learn More
Zeiss™ Digital Camera Adapter P95 M37/52x0.75 for Primo Star™ and Primo Vert™ Microscopes
Click to view available options
For Use With (Equipment):
Primo Star and Primo Vert Microscopes
Unit Size:
Each
Description
For use with Zeiss Microscopes
Specifications
Specifications
| Type | Digital Camera Adapter |
| Description | Accepts cameras with filter threads of M37mm or M52mm with a pitch of 0.75mm |
| For Use With (Equipment) | Primo Star and Primo Vert Microscopes |