Learn More
Carl Zeiss™ Microscope Stage Micrometers
Finely divided stage micrometer for calibrating optical systems with eyepiece graticule patterns. Carl Zeiss™ Microscope Stage Micrometer includes 1mm and 1/100mm intervals.
475.94€ - 480.18€
Specifications
Type | Stage Micrometer |
---|---|
Includes | Stage micrometer, 5 lines with 1mm intervals in y-direction |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11884953
|
Carl Zeiss™
474026-0000-000 |
Transmitted light microscope, eyepiece graticule |
480.18€
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
11866422
|
Carl Zeiss™
474027-0000-000 |
Reflected light microscope, eyepiece graticule |
475.94€
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
Description
- For calibrating optical systems with eyepiece graticule patterns
- 5 lines with 1mm intervals in y-direction
- 100 lines with 1/100mm intervals in y-direction
Specifications
Stage Micrometer | |
Stage micrometer, 5 lines with 1mm intervals in y-direction |