Learn More
Carl Zeiss™ Replacement Bulbs for Microscopes and Fiber Optic Power Supplies
Click to view available options
Type:
Halogen Bulb
Halogen Reflector Bulb
Mercury HBO
Wattage:
20 w
103 w
150 w
For Use With (Equipment):
Axio Examiner, Imager, Observer, Scope; Axioplan; Axioskop™; Axiotech; Axiovert™; PALM; Stemi; and SteREO
Axiostar Plus Student, Axiostar Plus ICS
ID03 Inverted Microscope
KL1500 Fiber Optic Power Supply
KL200 Fiber Optic Power Supply
Unit Size:
Each
Description
- Accessory for use with Carl Zeiss Stereomicrscopes
- Halogen bulb
Specifications
Specifications
| Type | Halogen Bulb |
| For Use With (Equipment) | Axiostar Plus Student, Axiostar Plus ICS |
| Length (English) | 1.18 in. |
| Length (Metric) | 29.9 mm |
| Voltage | 6 V |
| Wattage | 20 w |