Learn More
Veolia Water Systems™ Cartidges
Product Code. 11960457
Click to view available options
Model:
LC182
LC208
Unit Size:
Each
Product Code. 11960457
Brand: Veolia Water Systems™ LC182
Only null left
Add to basket
Add to basket
Robust and reliable water purification cartridges ideal for removing ionic contaminants to achieve specified water quality.
  • Purification cartridges
  • Removes ionic contaminants to achieve specified water quality
  • Compatible with PURELAB Ultra Mk1, PURELAB Ultra Mk2
Specifications
Model LC182