Learn More
Epredia™ Cassette 1 Slotted Tissue Cassettes
Product Code. 15327260 Shop All Epredia Products
Click to view available options
Color:
Blue
Unit Size:
Pack of 500
Product Code. 15327260
Brand: Epredia™ 1000961
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Hold tissue specimens during processing and embedding with Epredia™ Cassette 1 slotted tissue cassettes.
  • Acetal polymer construction
  • 30° writing face
  • Small slot-style pores
Ordering Alerts:
  • Supplied in box of 500 cassettes with attached breakaway lids (not available in Tube Packs)
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Slotted Pore
Writing Face Angle 30°
Color Blue
Material Acetal Polymer
Product Type Tissue Cassette