Learn More
Memmert™ Castor Frame (2-Part) for UN30
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN30 Oven
Unit Size:
Each
Description
- 2-parts
- Height: 140 mm
Specifications
Specifications
| For Use With (Equipment) | UN30 Oven |
| Type | Castor Frame |
| Height (Metric) | 140 mm |