Learn More
CCF4-AM
Product Code. 11533147
Click to view available options
Quantity:
5mg
20mg
Unit Size:
20mg
5mg
Product Code. 11533147
Brand: Invitrogen K1028
CCF4-AM
CCF4-AM, our LiveBLAzer™ substrate, is a lipophilic, esterified form of the CCF4 substrate which allows it to readily enter cells. Upon entry, cleavage by endogenous cytoplasmic esterases rapidly converts CCF4-AM into its negatively charged form, CCF4, which is retained in the cytosol. CCF4 is a Fluorescence Resonance Energy Transfer (FRET) substrate which consists of a cephalosporin core linking 7-hydroxycoumarin to fluorescein. This product is to be used as a FRET-based substrate for β-lactamase as a sensitive reporter of mammalian gene expression in the GeneBLAzer™ technology.
TRUSTED_SUSTAINABILITY
Specifications
Cell Permeability Cell-Permeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Solid
Product Line GeneBLAzer
Quantity 5mg
Substrate CCF4-AM
Substrate Properties Chemical Substrate
Substrate Type Beta-Lactamase Substrate
For Use With (Equipment) Flow Cytometer, Microplate Reader
Shipping Condition Room Temperature