Learn More
Motic™ CCIS™ Plan Achromatic Objective UC
Color corrected infinity optical system/plan achromat objective for crisp, flat, high-contrast images.
Brand:  Motic™ 1101001705251
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15970571
  • 173.00€ / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
  • Color corrected infinity system.
    • Brilliant, flawless image quality.
    • Image aberrations are corrected right to the edge of the field of view.
  • Plan achromat lenses.
    • Provide field flatness.
  • Suitable for the following:
    • Panthera U Binocular
    • Panthera U Trinocular
    • Panthera KU Binocular
    • Panthera KU Trinocular
    • Panthera C Binocular
    • Panthera C Trinocular
    • Panthera CC Binocular
    • Panthera CC Trinocular
    • Panthera DL
    • Panthera L
Specifications
Specifications
Objective type: Plan achromatic
Objective NA: 1.25, WD: 0.16 mm, DIN: 45 mm
Objective tube length: Infinity CCIS™
Objective mounting thread: W 4/5 in x 1/36 in. (RMS standard)
Panthera series microscopes
100X