Learn More
Motic™ CCIS™ EC Plan Achromatic Objective
Description
- EC Plan achromatic objective
- Incorporates Color Corrected Infinity Optics: CCIS™
- Specifications: 60X/0.80/S (WD=0.35mm)
- For use with BA Series Microscopes
Order Info
MagnificationPower
Specifications
Specifications
| Type | CCIS, Plan Achromatic Phase Objective |
| Field of View | EC Plan |
| Focus Distance | 0.35 WD |
| For Use With (Equipment) | BA Microscope Series |
| Magnification Power | 60X |
| Numerical Aperture | 0.80 |
| Quantity | 1 Ea. |