Learn More
CD105 (Endoglin) Monoclonal Antibody (MJ7/18), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
68.25€ - 295.00€
Specifications
Antigen | CD105 (Endoglin) |
---|---|
Clone | MJ7/18 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15297147
|
Affymetrix eBioscience
14-1051-82 |
100 μg |
118.00€
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15207157
|
Affymetrix eBioscience
14-1051-85 |
500 μg |
295.00€
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15287147
|
Affymetrix eBioscience
14-1051-81 |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
Description: The MJ7/18 monoclonal antibody reacts with the mouse CD105 molecule, also known as Endoglin. This 90 kDa disulfide-linked homodimer is expressed by vascular endothelial cells. It is suggested that CD105 functions in adhesion and embryonic angiogenesis. Applications Reported: The MJ7/18 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and immunohistochemical staining of frozen tissue sections. Applications Tested: The MJ7/18 antibody has been tested by flow cytometric analysis of bEnd.3 cells or mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.Specifications
CD105 (Endoglin) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
ENG | |
Primary | |
4° C | |
ENG |
MJ7/18 | |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q63961 | |
13805 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.