missing translation for 'onlineSavingsMsg'
Learn More

CD107b (LAMP-2) Monoclonal Antibody (eBioABL-93 (ABL-93)), eFluor™ 660, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-1072-82

Additional Details : Weight : 0.25000kg

Product Code. 15361880

  • 252.00€ / 100µg
Estimated Shipment: 06-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The eBioABL-93 monoclonal antibody reacts with mouse CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated protein of approximately 120 kDa that is expressed intracellularly in lysosomal membranes. It is also transiently expressed on the surface of cytolytic T cells during degranulation, but to a lesser extent than CD107a. It has been reported that CD107b may be identical to the mouse Mac-3 antigen. Applications Reported: This ABL-93 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This eBioABL-93 (ABL-93) antibody has been tested by intracellular staining and flow cytometric analysis of thioglycolate elicited peritoneal mouse cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD107b (LAMP-2) Monoclonal antibody specifically detects CD107b (LAMP-2) in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD107b (LAMP-2)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P17047
LAMP2
Affinity chromatography
RUO
16784
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
eBioABL-93 (ABL-93)
eFluor 660
LAMP2
CD107 antigen-like family member B; CD107b; EGK_20887; Igp96; Lamp II; LAMP2; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; Lamp-2a; Lamp-2b; Lamp-2c; LAMPB; LGP110; LGP-110; LGP-96; LGP-B; lysosomal associated membrane protein 2; lysosomal membrane glycoprotein 2; lysosomal membrane glycoprotein type B; lysosomal-associated membrane protein 2; lysosome-associated membrane glycoprotein 2; lysosome-associated membrane protein 2; Mac3; RP23-193O17.2
Rat
100 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only.