missing translation for 'onlineSavingsMsg'
Learn More

CD127 / IL-7Ra Rat, Biotin, Clone: A7R34, enQuireBio™

Rat Monoclonal Antibody

85.85€ - 364.00€

Specifications

Antigen CD127 / IL-7Ra
Clone A7R34
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15922198
View Documents Promotion Details
EnquireBio
QAB55-B-25UG
25 μg
85.85€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15912198
View Documents Promotion Details
EnquireBio
QAB55-B-100UG
100 μg
250.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15932198
View Documents Promotion Details
EnquireBio
QAB55-B-500UG
500 μg
364.00€
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD127 / IL-7Ra
0.5mg/mL
Monoclonal
Rat
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
16172
IgG2a κ
Protein A/G Purified
A7R34
Flow Cytometry
Biotin
RUO
NP_032385
Il17ra
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications