missing translation for 'onlineSavingsMsg'
Learn More

CD15 Ab-3 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry procedures with Epredia™ CD15 Ab-3, Mouse Monoclonal Antibody.

134.00€ - 400.00€

Specifications

Antigen CD15 Ab-3
Clone MMA; same as LeuM1
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Product Code Brand Form Concentration Quantity Price Quantity & Availability  
Product Code Brand Form Concentration Quantity Price Quantity & Availability  
12689396
View Documents Promotion Details
Epredia
MS-1259-P0
Concentrated 200μg/mL 100 μL
134.00€
100µL
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12699396
View Documents Promotion Details
Epredia
MS-1259-P1
Concentrated 200μg/mL 500 μL
264.00€
500µL
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12679396
View Documents Promotion Details
Epredia
MS-1259-P
Concentrated 200μg/mL 1 mL
370.00€
1mL
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12649406
View Documents Promotion Details
Epredia
MS-1259-R7
Prediluted 7 mL
264.00€
7mL
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12659406
View Documents Promotion Details
Epredia
MS-1259-RQ
Ready-to-Use 12 mL
400.00€
12mL
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12609406
View Documents Promotion Details
Epredia
MS-1259-P1ABX
Concentrated 1mg/mL 100 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12619406
View Documents Promotion Details
Epredia
MS-1259-PABX
Concentrated 1mg/mL 200 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

3-fucosyl-N-acetyllactosamine (3-FAL) or CD15 or X-hapten plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. CD15 is also expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin's disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Host Species: Mouse

Clone: MMA; same as LeuM1

Isotype: IgM/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: U937 histiocytic cell line

Molecular Weight: 220kDa

Positive Control: Reed-Sternberg's cells in Hodgkin's lymphoma

Cellular Localization: Membrane and granular paranuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Specifications

Specifications

CD15 Ab-3
Flow Cytometry, Immunohistochemistry (Paraffin)
Unconjugated
IVD
U937 histiocytic cell line
MMA; same as LeuM1
Monoclonal
Mouse
Oncogenesis
Primary
Videos
SDS
Documents

Documents

Product Certifications