missing translation for 'onlineSavingsMsg'
Learn More

CD19 Rat, APC, Clone: 1D3, enQuireBio™

Rat Monoclonal Antibody

98.45€ - 276.00€

Specifications

Antigen CD19
Clone 1D3
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15941758
View Documents Promotion Details
EnquireBio
QAB28-APC-25UG
25 μg
98.45€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15931758
View Documents Promotion Details
EnquireBio
QAB28-APC-100UG
100 μg
276.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD19
0.5mg/mL
Monoclonal
Rat
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
12478
IgG2a κ
Protein A/G Purified
1D3
Flow Cytometry
APC
RUO
NP_033974
Cd19
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications