Learn More
CD25/IL-2Rα Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunofluorescence and flow cytometry experiments with Epredia™ CD25/IL-2Rα Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-203-P1
Additional Details : Weight : 0.00190kg
Description
IL-2R (T-cell growth factor receptor) consists of a chain of 55kDa (also known as Tac antigen or p55 or α chain) and a p75 or Β chain. IL-2Rα is over-expressed on the surface of activated T-cells, HTLV-1 or -2 positive T-cell lines, and moderately expressed on activated B-cells, NK-cells, and macrophages.Host Species: Mouse
Clone: IL2R.1
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant human interleukin-2 receptor
Molecular Weight: 55 kDa
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
CD25/IL-2Ralpha Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
B-Cell | |
Human | |
IgG2a κ |
Flow Cytometry, Immunofluorescence | |
IL2R.1 | |
Unconjugated | |
Recombinant human interleukin-2 receptor | |
RUO | |
Primary | |
Concentrated |