Learn More
CD314 (NKG2D) Monoclonal Antibody (CX5), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
198.00€ - 338.00€
Specifications
Antigen | CD314 (NKG2D) |
---|---|
Clone | CX5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15370980
|
Affymetrix eBioscience
25-5882-81 |
50 μg |
198.00€
50µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15380980
|
Affymetrix eBioscience
25-5882-82 |
100 μg |
338.00€
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The CX5 monoclonal antibody reacts with the mouse NKG2D, a lectin-like molecule expressed on both human and mouse NK cells. Mouse NKG2D binds to retinoic acid-inducible RAE-1 alpha, beta, gamma, delta, epsilon and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors, through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1^+ thymocytes, in vitro activated LAK cells, and a subset of splenic NKT cells. Applications Reported: This CX5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CX5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD314 (NKG2D) Monoclonal antibody specifically detects CD314 (NKG2D) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD314 (NKG2D) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD314; D12S2489E; D6H12S2489E; killer cell lectin like receptor K1; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; KLR; Klrk1; natural killer cell group 2D; natural killer cell receptor; NK cell receptor D; NK lectin-like receptor; Nkg2d; NKG2-D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor; NKLLR; Nkrp2; NKR-P2; NKR-P2, ortholog of human NKG2D | |
Klrk1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Klrk1 |
CX5 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
O54709 | |
27007 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.