missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35/CR1 (Follicular Dendritic Cell Marker) Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments.
142.00€ - 566.10€
Specifications
Antigen | CD35 |
---|---|
Clone | E11 |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
12661037
|
Epredia
MS-336-P0 |
Concentrated | 800μg/mL | 100 μL |
142.00€
100µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12671037
|
Epredia
MS-336-P1 |
Concentrated | 800μg/mL | 500 μL |
391.00€
500µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12621047
|
Epredia
MS-336-R7 |
Prediluted | — | 7 mL |
204.00€
7mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12681037
|
Epredia
MS-336-P1ABX |
Concentrated | 1mg/mL | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12691037
|
Epredia
MS-336-PABX |
Concentrated | 1mg/mL | 200 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12651037
|
Epredia
MS-336-P |
Concentrated | 800μg/mL | 1 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
Description
Complement receptor 1 (CR1, CD35) is a cell membrane-bound, monomeric glycoprotein that is found on numerous cell types of primates including erythrocytes, lymphocytes, and phagocytes. The primary function of CR1 is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules.Host Species: Mouse
Clone: E11
Isotype: IgG1
Species Reactivity: Human, Rhesus Monkey and Baboon. Others not tested.
Epitope: Distal from the C3b-binding site
Immunogen: Intact human monocytes
Molecular Weight: 210-220 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
CD35 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Non-human Primate | |
IgG1 |
E11 | |
Monoclonal | |
Mouse | |
Hematopoietic | |
Intact human monocytes | |
Primary |