Learn More
Thermo Scientific™ CD36 Peptide P (93-110)
For cellular and molecular biology applications
Brand:  Thermo Scientific™ J66970.|D
Additional Details : Weight : 0.00001kg
 View more versions of this product
Product Code. 15408559
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
HPLC
Lyophilized Powder
CD36 Peptide P (93-110)
RUO - research use only
-30°C to -10°C
2270.45
0.5 mg
Tyr-Arg-Val-Arg-Phe-Leu-Ala-Lys-Glu-Asn-Val-Thr-Gln-Asp-Ala-Glu-Asp-Asn-Cys
Product Suggestions
Product Suggestions
Videos
SDS Documents