missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences CD4 Isolation Kit for FABian™

Product Code. 15859774 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Kit, Human
10 Kits, Human
Unit Size:
1 set
10 sets
This item is not returnable. View return policy

Product Code. 15859774

Brand: IBA Lifesciences 66002010

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

IBA Lifesciences™ CD4 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.

  • The kit is used for the selection of CD4+ T cells from whole blood or buffy coat in conjunction with the FABian™ system.
  • It includes CD4 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian™ run to isolate cells from 9 ml whole blood.

Specifications

Product Type Non-magnetic Cell Isolation Kit
Content And Storage 2°C to 8°C
Donor Source Human Whole Blood
For Use With (Application) Isolation of label-free cells
Sufficient For 9 mL Whole Blood
Quantity 10 Kits, Human
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.