missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse, V450, Clone: OKT4, enQuireBio™
Mouse Monoclonal Antibody
124.00€ - 250.00€
Specifications
Antigen | CD4 |
---|---|
Clone | OKT4 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931508
|
EnquireBio
QAB11-V450-25TESTS |
25 Tests |
124.00€
25 tests |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921508
|
EnquireBio
QAB11-V450-100TESTS |
100 Tests |
250.00€
100 tests |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD4 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
920 | |
IgG2b κ | |
Protein A/G Purified |
OKT4 | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
NP_001181945 | |
CD4 | |
Primary | |
Store at 2-4°C. |