missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40 / TNFRSF5 Mouse, Clone: C40/1605, enQuireBio™
Mouse Monoclonal Antibody
199.00€ - 510.00€
Specifications
Antigen | CD40 / TNFRSF5 |
---|---|
Clone | C40/1605 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15921248
|
EnquireBio
958-MSM1-P1 |
100 μg |
510.00€
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911248
|
EnquireBio
958-MSM1-P0 |
20 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
CD40 / TNFRSF5 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
958 | |
Recombinant human full-length CD40 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
43kDa |
C40/1605 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_016883624 | |
CD40 | |
IgG1 κ | |
Protein A/G Purified | |
Cell Surface and Secreted | |
L-428 Cells. Tonsil, Spleen or Thymus |