Learn More
CD42b, FITC, clone: MB45, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD42B01
Additional Details : Weight : 0.02100kg
Description
Single-chain membrane glycoprotein that forms a non-covalent complex with GPIb. (MW 23kDa) Reactivity with resting and activated platelets, weakly on monocytes, megakaryocytes and attachment site for the platelet plasma membrane to the submembrane cytoskeleton. GPIb/IX complex, functions as the receptor for ristocetin-induced binding of von Willebrand factor and as the von Willebrand factor-depend adhesion receptor.
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.Specifications
CD42b | |
Monoclonal | |
FITC | |
GP1BA | |
CD42a, GPIX, GP-IX, glycoprotein 9, platelet glycoprotein IX, Platelet GPIX | |
Mouse | |
Purified | |
ASR, ASR/IVD | |
2811 | |
Store at 2 - 8 °C. | |
Liquid |
Flow Cytometry | |
MB45 | |
PBS with 10% sucrose, 4mg/mL BSA and 0.1% sodium azide | |
P07359 | |
GP1BA | |
Human CD42b | |
0.5 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |