missing translation for 'onlineSavingsMsg'
Learn More

CD45.1 Mouse, APC, Clone: A20, enQuireBio™

Mouse Monoclonal Antibody

124.00€ - 276.00€

Specifications

Antigen CD45.1
Clone A20
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15942008
View Documents Promotion Details
EnquireBio
QAB42-APC-25UG
25 μg
124.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15932008
View Documents Promotion Details
EnquireBio
QAB42-APC-100UG
100 μg
276.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD45.1
0.5mg/mL
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
19264
IgG2a κ
Protein A/G Purified
A20
Flow Cytometry
APC
RUO
NP_001255215
Ptprc
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications