missing translation for 'onlineSavingsMsg'
Learn More

EnquireBio CD45.1 Mouse, APC, Clone: A20, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB42-APC-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932008

  • 247.00€ / 100µg

Estimated Shipment: 02-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

CD45.1
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Ptprc
Protein A/G Purified
RUO
19264
IgG2a κ
Flow Cytometry
A20
APC
NP_001255215
Mouse
100 μg
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions