missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio CD45.2 Mouse, PE-Cy7, Clone: 104, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB43-PE7-25ug
Additional Details : Weight : 0.01000kg
Specifications
CD45.2 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Ptprc | |
Protein A/G Purified | |
RUO | |
19264 | |
IgG2a κ |
Flow Cytometry | |
104 | |
PE-Cyanine7 | |
NP_001255215 | |
Mouse | |
25 μg | |
Primary | |
Store at 2-4°C. |