missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Rat, Biotin, Clone: 30-F11, enQuireBio™
Rat Monoclonal Antibody
85.85€ - 137.00€
Specifications
Antigen | CD45 |
---|---|
Clone | 30-F11 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15921968
|
EnquireBio
QAB40-B-25UG |
25 μg |
85.85€
25µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911968
|
EnquireBio
QAB40-B-100UG |
100 μg |
137.00€
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD45 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2 | |
19264 | |
IgG2b κ | |
Protein A/G Purified |
30-F11 | |
Flow Cytometry | |
Biotin | |
RUO | |
NP_001255215 | |
Ptprc | |
Primary | |
Store at 2-4°C. |