missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD62L / L-Selectin Rat, Qfluor 710, Clone: MEL-14, enQuireBio™
Rat Monoclonal Antibody
200.00€ - 263.00€
Specifications
Antigen | CD62L / L-Selectin |
---|---|
Clone | MEL-14 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932128
|
EnquireBio
QAB49-QF710-25UG |
25 μg |
200.00€
25µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922128
|
EnquireBio
QAB49-QF710-100UG |
100 μg |
263.00€
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD62L / L-Selectin | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2 | |
20343 | |
IgG2a κ | |
Protein A/G Purified |
MEL-14 | |
Flow Cytometry | |
Qfluor 710 | |
RUO | |
NP_001157531 | |
Sell | |
Primary | |
Store at 2-4°C. |