missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ CDX2 Rabbit Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CDX2, Rabbit Monoclonal Antibody.
260.00€ - 1375.00€
Specifications
Antigen | anti-CDX2 |
---|---|
Clone | EPR2764Y |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12623657
|
Epredia™
RM-2116-R7 |
Ready-to-Use | 7 mL |
415.00€
7mL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12613657
|
Epredia™
RM-2116-RQ |
Ready-to-Use | 12 mL |
645.00€
12mL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12653657
|
Epredia™
RM-2116-S0 |
Supernatant | 100 μL |
260.00€
100µL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12643657
|
Epredia™
RM-2116-S |
Supernatant | 1 mL |
1375.00€
1mL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The caudal-related homeodomain protein2 (CDX2) is a transcription factor shown to play a role in the development of small and large intestine in mammals and in the differentiation of intestinal epithelial cells. It has been shown that CDX2 protein detection by IHC coorelates with RNA transcript levels. In studies using 745 cancers from many anatomic sites, colonic adenocarcinomas demonstrated strong staining in 90% of the cases, while adenocarcinomas of the stomach, esophagus and ovary showed extensive staining in only 30% of the cases. Stains of CDX2 and Villin are useful markers in differntial diagnosis of bladder adenocarcinoma and secondary colorectal adenocarcinoma.Host Species: Rabbit
Clone: EPR2764Y
Species Reactivity: Human. Others not tested.
Ig Isotype: Rabbit IgG
Epitope: not determined
Immunogen: A synthetic peptide corresponding to residues near the N-terminus of human CDX2
Molecular Weight: 40 kDa
Positive Control: Colon carcinomas
Cellular Localization: Nuclear with occasional cytoplasmic staining on colon carcinoma
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
anti-CDX2 | |
Immunohistochemistry | |
Unconjugated | |
A synthetic peptide corresponding to residues near the N-terminus of human CDX2 |
EPR2764Y | |
Monoclonal | |
Rabbit | |
Primary |