missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CE3F4

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CE3F4 is a noncompetitive Epac1 inhibitor. Blocks Epac-induced Rap activation and prevents isoprenaline -induced autophagy flux in cardiomyocytes. Has no effect on PKA activity in the presence of cAMP.
Specifications
Specifications
| Purity | >98% |
| Quantity | 10 mg |
| Product Type | CE3F4 |
| Molecular Weight (g/mol) | 351.01 |
| Inhibitors | EPAC Inhibitors |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction