missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cefoxitin sodium salt, 94%

Product Code. 15545136 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15545136

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cefoxitin sodium salt, CAS # 33564-30-6, is a β-lactam, second-generation cephalosporin compound showing bactericidal activity against Bacteroides, gram-negative bacteria, including strains of P. mirabilis, P. vulgaris, E. coli, S. typhimurium, and S. enteritidis. | CAS: 33564-30-6 | C16H16N3NaO7S2 | 449.428 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cefoxitin sodium salt is resistant to β-lactamase
  • It binds and inactivates penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall thus interfering with the cross-linkage of peptidoglycan chains necessary for bacterial cell wall strength and rigidity

Applications

  • Cefoxitin sodium salt is the sodium salt form of cefoxitin derived from cephamycin C
  • It is a β-lactam, second-generation cephalosporin compound with bactericidal activity against numerous species such as Bacteroides, gram-negative bacteria, including strains of P. mirabilis, P. vulgaris, E. coli, S. typhimurium, and S. enteritidis
  • Cefoxitin increases survival in mice infected with S. aureus, S. pyogenes, and K. pneumoniae
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33564-30-6
Molecular Formula C16H16N3NaO7S2
Molecular Weight (g/mol) 449.428
InChI Key GNWUOVJNSFPWDD-XMZRARIVSA-M
PubChem CID 23667300
ChEBI CHEBI:3501
IUPAC Name sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]

Specifications

Type Cefoxitin Sodium Salt
Infrared Spectrum Authentic
Specific Rotation +206° to +214° (20°C, 589nm) (c= 1, methanol)
Water 1% max.
Quantity 1 g
Formula Weight 449.43
Percent Purity 94%
Chemical Name or Material Cefoxitin sodium salt
compliance-icons
Product Identifier
  • Cefoxitin sodium salt
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.