missing translation for 'onlineSavingsMsg'
Learn More

Ceftazidime pentahydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7112897
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
78439-06-2
C22H22N6O7S2·5H2O
636.65
NMVPEQXCMGEDNH-TZVUEUGBSA-N
131673973
CHEBI:3509
2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate
CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O
This item is not returnable. View return policy
78439-06-2
C22H22N6O7S2·5H2O
636.65
NMVPEQXCMGEDNH-TZVUEUGBSA-N
131673973
CHEBI:3509
2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate
CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >150.0°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Merck Index 13,1959
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alkali,slightly soluble in methanol,dmf,insoluble in acetone,ethanol,chloroform,ether,,ethyl acetate,toluene,dioxane
Formula Weight 636.65
Percent Purity 98%
Chemical Name or Material Ceftazidime pentahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'